Learn More
Waters Corp Plunger Seal, Tan (Rulon)
Catalog No. 50793528 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The tan Plunger Seal is generally used in systems that run GPC solvents (THF, TCB).
Catalog No. 50-793-528 Supplier Waters Corp Supplier No. WAT025384
The tan Plunger Seal is generally used in systems that run GPC solvents (THF, TCB).