Learn More
Waters Corp Plunger Seals Replacement Kit (Std-Yellow) 2/pk
Catalog No. NC9563013 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed specifically for Waters Alliance Solvent Management Systems, these standard plunger seals are made from a Waters proprietary material and are the best choice for most applications.
Catalog No. NC9563013 Supplier Waters Corp Supplier No. WAT270938
Designed specifically for Waters Alliance Solvent Management Systems, these standard plunger seals are made from a Waters proprietary material and are the best choice for most applications.