Learn More
Waters Corp Waters Plunger Seals Kit, Clear, 2/pk, HPLC
Catalog No. 50792551 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These optional (clear) plunger seals for Waters™ Alliance Solvent Management Systems are good alternatives if the standard seal does not perform well for your specific application.
  • Clear color provides an optional alternative to standard seals.
  • Contains 2 seals per package.
  • Designed for HPLC systems.
  • Replace as required for optimal performance.
  • Compatible with Waters™ Alliance Solvent Management Systems.
Catalog No. 50-792-551 Supplier Waters Corp Supplier No. 700001326
These optional (clear) plunger seals for Waters™ Alliance Solvent Management Systems are good alternatives if the standard seal does not perform well for your specific application.
  • Clear color provides an optional alternative to standard seals.
  • Contains 2 seals per package.
  • Designed for HPLC systems.
  • Replace as required for optimal performance.
  • Compatible with Waters™ Alliance Solvent Management Systems.