Learn More
Waters Corp Waters Plunger Seals Replacement Kit, Black, 2/pk, HPLC
Catalog No. 502039981 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These optional black plunger seals are designed for Alliance Solvent Management Systems. They serve as alternatives if the standard seal does not perform well for a specific application.
  • Seal type
  • Designed for HPLC system applications
  • Black color
  • Package contains 2 seals
  • Replace as required
  • Serve as alternatives if standard seals do not perform well for specific applications
Catalog No. 50-203-9981 Supplier Waters Corp Supplier No. WAT271066
These optional black plunger seals are designed for Alliance Solvent Management Systems. They serve as alternatives if the standard seal does not perform well for a specific application.
  • Seal type
  • Designed for HPLC system applications
  • Black color
  • Package contains 2 seals
  • Replace as required
  • Serve as alternatives if standard seals do not perform well for specific applications