Learn More
Ace Glass, Inc. PLUNGER VALVE15 3/8IN TUBE END
SDP
Catalog No. NC3584884
Encompass_Preferred
Plunger valve,#15,3/8in tube end.Ace Glass pressure tubes and bottles are the premier glass pressure vessels in the industry. Flat-bottomed bottles are rated at 60psi at room temperature while our tubes are rated at 150psi at as much as 120 degrees C.
Catalog No. NC3584884 Supplier Ace Glass, Inc. Supplier No. 864820
Plunger valve,#15,3/8in tube end.Ace Glass pressure tubes and bottles are the premier glass pressure vessels in the industry. Flat-bottomed bottles are rated at 60psi at room temperature while our tubes are rated at 150psi at as much as 120 degrees C.