Learn More
Electron Microscopy Sciences Plunging Tweezers (CP3)
SDP
Catalog No. 501926621
Encompass_Preferred
Custom tweezers specifically made to fit the Gatan Cryoplunge an instrument used in the preparation of frozen hydrated specimens for cryoEM.
Catalog No. 50-192-6621 Supplier Electron Microscopy Sciences Supplier No. CP3690
Custom tweezers specifically made to fit the Gatan Cryoplunge an instrument used in the preparation of frozen hydrated specimens for cryoEM.