Learn More
Ace Glass, Inc. Stopper, cone, 37mm OD top, 22mm OD bottom, black neoprene, pk12
SDP
Catalog No. NC9419231
Encompass
22mm to 37mm OD black neoprene nested ring pluro stoppers. Will give a vacuum tight fit for all openings within its size range. Suitable for applications from -40 to 150 degrees F.
Catalog No. NC9419231 Supplier Ace Glass, Inc. Supplier No. 1201446
22mm to 37mm OD black neoprene nested ring pluro stoppers. Will give a vacuum tight fit for all openings within its size range. Suitable for applications from -40 to 150 degrees F.