Learn More
Pluriselect pluriStrainer (Cell Strainer) 15 um mesh-size PET, single packed, sterile, 25/pk
Supplier:  Pluriselect 435001503
Encompass
The Flip-Strainer Brand “Pluristrainer” is a separation device (or cell strainer) for a wide range of laboratory applications where the filtration, particle separation and the purification of liquids is required. Its unique design features improved ventilation to avoid clogging for a smart sample preparation. Pluristrainers are stackable to a cascade of strainers. They are flip-strainers and allow flashing back the separated particles. For large sample volumes it can be combined with a funnel. It fits all major brand 50 mL centrifuge . (conical) tubes. The combination with the Connector Ring allows the evacuation of the 50 ml tube-Strainer unit to increase the flow rate. Pressure exchange can be completely blocked. The sample will now stay inside the strainer and can be incubated/treated, including with commonly used cell lysis reagents. This product line cannot be autoclaved!
Catalog No. NC1203375