Learn More
ATCC Pluralibacter gergoviae
Catalog No. 502382798
Encompass
Pluralibacter gergoviae strain CDC 604-77 [CIP 76.01] was isolated from urine in France. This whole-genome sequenced bacterial type strain can be used in assays of antimicrobial preservatives.
Catalog No. 50-238-2798 Supplier ATCC Supplier No. 33028
Pluralibacter gergoviae strain CDC 604-77 [CIP 76.01] was isolated from urine in France. This whole-genome sequenced bacterial type strain can be used in assays of antimicrobial preservatives.