Learn More
Teknova PluriFreeze™ Base, 100mL, Sterile.
SDP
Catalog No. 502995672
Encompass
PluriFreeze™ Base is a wash that mimics intracellular space and provides metabolic support to cells. Plurifreeze base can be used across the cryopreservation workflow.
Catalog No. 50-299-5672 Supplier Teknova Supplier No. 22244
PluriFreeze™ Base is a wash that mimics intracellular space and provides metabolic support to cells. Plurifreeze base can be used across the cryopreservation workflow.