Learn More
Teknova PluriFreeze™ PF10, 100 mL, Sterile.
SDP
Catalog No. 502995671
Encompass
PluriFreeze™ PF10 is a low viscosity freezing medium with 10% DMSO that simplifies scale-up and process automation.
Catalog No. 50-299-5671 Supplier Teknova Supplier No. 22243
PluriFreeze™ PF10 is a low viscosity freezing medium with 10% DMSO that simplifies scale-up and process automation.