Learn More
Pluriselect PluriMate II 50 ml tubes, unfilled , 100 Pc
Catalog No. NC1229225
Encompass
pluriMate was developed for optimal separation of leukocytes and peripheral blood mononuclear cells (PBMC) from whole blood and bone marrow. The key feature of pluriMate is the mesh supported barrier incorporated at the bottom of the centrifuge tube. It prevents you from time-consuming and laborious overlaying of the sample material. Anticoagulated blood or bone marrow can simply be poured directly from the blood sampling tube into the pluriMate tube. The barrier prevents mixture of the sample material with the separation medium. During the centrifugation the white blood cells (on the basis of their density) are separated depending on the used density gradient medium (Leuko Spin, PBMC Spin, PLT Spin etc.) and will be enriched above the mesh supported barrier and the separation medium. When the separation is complete, the barrier prevents a contamination of the enriched cell fraction during harvest with unwanted cells.
Catalog No. NC1229225 Supplier Pluriselect Supplier No. 441005011
pluriMate was developed for optimal separation of leukocytes and peripheral blood mononuclear cells (PBMC) from whole blood and bone marrow. The key feature of pluriMate is the mesh supported barrier incorporated at the bottom of the centrifuge tube. It prevents you from time-consuming and laborious overlaying of the sample material. Anticoagulated blood or bone marrow can simply be poured directly from the blood sampling tube into the pluriMate tube. The barrier prevents mixture of the sample material with the separation medium. During the centrifugation the white blood cells (on the basis of their density) are separated depending on the used density gradient medium (Leuko Spin, PBMC Spin, PLT Spin etc.) and will be enriched above the mesh supported barrier and the separation medium. When the separation is complete, the barrier prevents a contamination of the enriched cell fraction during harvest with unwanted cells.