Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pluripotent Stem Cell Immunocytochemistry Kit (OCT4, SSEA4)

Catalog No. A25526
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25526 40 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25526 Supplier Invitrogen™ Supplier No. A25526
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Pluripotent Stem Cell Immunocytochemistry Kit (OCT4, SSEA4) enables optimal image-based analysis of two key markers of human pluripotent stem cells (PSCs): OCT4 and SSEA4.

The Pluripotent Stem Cell Immunocytochemistry Kit (OCT4, SSEA4) enables optimal image-based analysis of two key markers of human pluripotent stem cells (PSCs): OCT4 and SSEA4. This high performance immunocytochemistry (ICC) kit includes a complete set of primary and secondary antibodies, a nuclear DNA stain, and premade buffers for an optimized staining experiment. To characterize PSCs with other marker combinations, we recommend the use of the Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit (Cat. No. A24881) or the Pluripotent Stem Cell Immunocytochemistry Kit (SOX2, TRA-1-60) (Cat. No. A25525).

With the Pluripotent Stem Cell Immunocytochemistry Kit (OCT4, SSEA4), you can:

  • Confirm expression of key pluripotent stem cell markers with highly specific antibodies
  • Get more information per sample by measuring more than one marker at a time
  • Generate beautiful multiplexed images with minimal effort
  • Save time and avoid cell loss by minimizing unnecessary wash steps in your staining protocol
  • Process samples with confidence using a complete and optimized set of ICC reagents

Confirm Expression of Key PSC Markers
The antibodies included in the kit have been carefully selected and validated for high performance in the ICC analysis of PSCs, whether cultured under feeder-dependent or feeder-free media systems. OCT4 is a critical nuclear marker, while SSEA4 is an important cell surface marker.

Get More Information Per Sample
The antibodies in the kit have been designed to allow for the specific and simultaneous assessment of OCT4 and SSEA4 along with nuclear DNA staining to help save time and precious sample.

Generate Beautiful Multiplexed Images
Powered by the superior performance of bright, photostable Alexa Fluor™ dyes, this kit enables the acquisition of beautiful multiplexed images of PSC marker expression using commonly available blue, green, and orange/red filter sets.

Minimize Unnecessary Wash Steps
The streamlined method recommended for this kit eliminates unnecessary wash steps found in standard ICC protocols to help save time and decrease the likelihood of cell loss during the staining protocol, especially when using glass coverslips or chamber slides. The only washes required are after incubation with primary and secondary antibodies.

Process Samples with Confidence
An optimized set of premade fixation, permeabilization, blocking, and wash buffers allows you to focus on answering scientific questions rather than spending time preparing reagents and determining compatibility.

What You Get
Easily scaled to any culture plate, dish, or slide format, the Pluripotent Stem Cell Immunocytochemistry Kit (OCT4, SSEA4) comes with reagents sufficient to stain 40 samples using a 50 uL staining volume. (See kit content details below.)

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594
Quantity 40 Tests
Content And Storage Store at -20°C to 4°C:
• Rabbit Anti-Human OCT4 (200X), 10 μL
• Mouse Anti-Human SSEA4 (100X), 20 μL
• Alexa Fluor™ 555 Donkey Anti-Rabbit (250X), 20 μL
• Alexa Fluor™ 594 Donkey Anti-Rabbit (250X), 20 μL
• Alexa Fluor™ 488 Goat Anti-Mouse IgG3 (250X), 20 μL

Store at -20°C to Room Temperature:
• NucBlue™ Fixed Cell ReadyProbes™ Reagent (1X), 1 vial
• Fixative Solution (1X), 10 mL
• Permeabilization Solution S (1X), 10 mL
• Blocking Solution (1X), 20 mL
• Wash Buffer (10X), 20 mL
Product Type Immunocytochemistry Kit
The Pluripotent Stem Cell Immunocytochemistry Kit provides the NucBlue Fixed Cell Stain. If I don't have the correct filter set for this stain, can I use another nucleic acid stain?

If you use Alexa Fluor 488/Alexa Fluor 555 dye-conjugated secondary antibodies and you have a Cy5/Alexa Fluor 647 filter set, you may use a far-red nucleic acid stain, such as NucRed Dead 647 ReadyProbes Reagent (Cat. No. R37113) or SYTOX Deep Red Nucleic Acid Stain (Cat. No. S11380). Do not use these far-red nucleic acid stains if using an Alexa Fluor 594 dye-conjugated secondary antibody as there is too much spectral overlap of Alexa Fluor 594 into the Cy5/Alexa Fluor 647 range.

Can I use the Pluripotent Stem Cell Immunocytochemistry Kit for flow cytometry analysis?

We do not recommend using the Pluripotent Stem Cell Immunocytochemistry Kit for flow cytometry analysis. The kit and protocol were specifically developed for imaging analysis (microscopy/high-content screening (HCS)).

Do you offer the individual components of the Pluripotent Stem Cell Immunocytochemistry Kit as standalone products?

Only the NucBlue Fixed Cell ReadyProbes Reagent may be purchased separately as Cat. No. R37606. We do offer the secondary antibodies separately, but not in the same proprietary concentration as provided in the kit. We also offer formaldehyde, DPBS, BSA, and saponin as separate catalog numbers, but not in the volumes or as formulated in the kit. The primary antibodies are not available as separate stock items; all non-stock items in the kit may be available as a custom order. Please contact your Account Representative.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.