Learn More
Medchemexpress LLC HY-10579 5mg Medchemexpress, Pluripotin CAS:839707-37-8 Purity:>98%
SDP
Catalog No. 501968294
Encompass_Preferred
Medchemexpress, HY-10579 5mg Pluripotin CAS:839707-37-8 Pluripotin is a dual inhibitor of ERK1 and RasGAP with KDs of 98 nM and 212 nM, respectively. Pluripotin also inhibits RSK1, RSK2, RSK3, and RSK4 with IC50s of 0.5, 2.5, 3.3, and 10.0 µM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8294 Supplier Medchemexpress LLC Supplier No. HY105795MG
Medchemexpress, HY-10579 5mg Pluripotin CAS:839707-37-8 Pluripotin is a dual inhibitor of ERK1 and RasGAP with KDs of 98 nM and 212 nM, respectively. Pluripotin also inhibits RSK1, RSK2, RSK3, and RSK4 with IC50s of 0.5, 2.5, 3.3, and 10.0 µM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.