Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PluriSTEM™ C3H/HeN Murine Embryonic Stem Cells

Catalog No. SCC055 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC055 2 vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC055 Supplier MilliporeSigma™ Supplier No. SCC055
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for pharmacological testing

  • Gender: Male
  • Karyotype: 40XY
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Place vials in liquid nitrogen immediately upon receipt until it is convenient to proceed to subculture.
Cell Type Murine Embryonic Stem Cells
For Use With (Application) Stem cell culture
Quantity 2 vials
Remarks Frozen in basal medium supplemented with 50% FCS and 10% DMSO

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.