Learn More
Pluriselect pluriStrainer Membrane-Mini (Cell Strainer) 8 um pore-size, 5 pcs. , sterile
Catalog No. NC1720798
Encompass_Preferred
Membrane filters have a well-defined pore size exclusion limit (available with 1, 3, 5 and 8 um pores), allowing for precise filtration and detection of very small particles. The membranes are non-hygroscopic (optimal for sample recovery), have low non-specific protein binding (reduces sample loss in protein-containing samples), are biologically inert, non-cytotoxic, and have excellent chemical and temperature resistance. The membrane filters can be used for example in biological or gravimetric analysis, cytology, parasitology or in environmental analyzes.
Catalog No. NC1720798 Supplier Pluriselect Supplier No. 461000805
Membrane filters have a well-defined pore size exclusion limit (available with 1, 3, 5 and 8 um pores), allowing for precise filtration and detection of very small particles. The membranes are non-hygroscopic (optimal for sample recovery), have low non-specific protein binding (reduces sample loss in protein-containing samples), are biologically inert, non-cytotoxic, and have excellent chemical and temperature resistance. The membrane filters can be used for example in biological or gravimetric analysis, cytology, parasitology or in environmental analyzes.