Learn More
Ace Glass, Inc. Stopper, cone, 46mm OD top, 29mm OD bottom, black neoprene, pk12
SDP
Catalog No. NC9757964
Encompass
29mm to 46mm OD black neoprene nested ring pluro stoppers. Will give a vacuum tight fit for all openings within its size range. Suitable for applications from -40 to 150 degrees F.
Catalog No. NC9757964 Supplier Ace Glass, Inc. Supplier No. 1201448
29mm to 46mm OD black neoprene nested ring pluro stoppers. Will give a vacuum tight fit for all openings within its size range. Suitable for applications from -40 to 150 degrees F.