Learn More
Biotium Pluronic F-127
Supplier:  Biotium 59000
Encompass_Preferred
Nonionic detergent useful for solubilizing large dye molecules such as Fura-2/AM, Fluo-3/AM, Indo-1/AM and Rhod-2/AM to facilitate cell loading. Pluronic F-127 is usually dissolved in DMSO or water with gentle heating. Properties: White solid soluble in water and DMSO; Store at room temperature; [9003-11-6]. Pluronic F-127 is a registered trade mark of BASF.
Catalog No. 50-196-4711