Learn More
Biotium Pluronic F-127, 10% in H2O
Catalog No. 501964712
Encompass_Preferred
Ready-to-use solution of Pluronic F-127, a nonionic detergent useful for solubilizing large dye molecules such as Fura-2/AM, Fluo-3/AM, Indo-1/AM and Rhod-2/AM to facilitate cell loading. Pluronic F-127 is a registered trade mark of BASF.
Catalog No. 50-196-4712 Supplier Biotium Supplier No. 59005
Ready-to-use solution of Pluronic F-127, a nonionic detergent useful for solubilizing large dye molecules such as Fura-2/AM, Fluo-3/AM, Indo-1/AM and Rhod-2/AM to facilitate cell loading. Pluronic F-127 is a registered trade mark of BASF.