Learn More
Abcam Pluronic F-127, 50G
Catalog No. 502654873
Encompass_Preferred
MW 102.13 Da. Pluronic F-127 is a non-ionic detergent useful for isolation of membrane proteins. It has been found to facilitate the solubilization of water-insoluble dyes and other materials in physiological media.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-4873 Supplier Abcam Supplier No. AB28541350G
MW 102.13 Da. Pluronic F-127 is a non-ionic detergent useful for isolation of membrane proteins. It has been found to facilitate the solubilization of water-insoluble dyes and other materials in physiological media.
The product is subject to the following: Abcam Restricted Use Statement