Learn More
Sigma Aldrich Fine Chemicals Biosciences Pluronic F-127, 9003-11-6, MFCD00082049, 1 KG
Catalog No. NC1588165
Encompass
Powder, BioReagent, suitable for cell culture. Pluronic F-127 was used to coat a siliconized coverslip to hold an egg extract in a study. Pluronic F-127 was added to phosphate buffered saline (PBS), to lower unspecific cell and protein adhesion to a PDMS-based microfluidic device.
Catalog No. NC1588165 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P24431KG
Powder, BioReagent, suitable for cell culture. Pluronic F-127 was used to coat a siliconized coverslip to hold an egg extract in a study. Pluronic F-127 was added to phosphate buffered saline (PBS), to lower unspecific cell and protein adhesion to a PDMS-based microfluidic device.