Learn More
Aat Bioquest Pluronic F-127 *20% solution in DMSO* | 9003-11-6 | 10mL
SDP
Catalog No. NC1086942
Encompass_Preferred
Pluronic F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells, and frequently used with dye AM esters to improve their water solubility. We recommend that it be stored at low temperature.
Catalog No. NC1086942 Supplier Aat Bioquest Supplier No. 20052
Pluronic F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells, and frequently used with dye AM esters to improve their water solubility. We recommend that it be stored at low temperature.