Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pluronic™ F-127 (20% Solution in DMSO)

Catalog No. P3000MP
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P3000MP 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P3000MP Supplier Invitrogen™ Supplier No. P3000MP
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Pluronic™ F-127 (20% Solution in DMSO)

Pluronic F-127 *20% solution in DMSO* is a nonionic, surfactant polyol (molecular weight approximately 12,500 daltons) that has been found to facilitate the solubilization of water-insoluble dyes and other materials in physiological media. Pluronic F-127 is commonly used to help disperse the acetoxymethyl (AM) esters of our ion indicators as well as our cell tracer dyes such as CFDA-SE.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Quantity 1 mL
I am using your FreeStyle Max 293 Expression System and my cell viability is at most 87-90%. Is this normal? Do you have any tips to increase the cell viability? I am using 125 mL and 250 mL non-baffled, polycarbonate shaker flasks with vented caps. Will using baffled flasks help? Should I add Pluronic reagent to the medium?

Culture viability should be in the high 90% range for the seed stock that you are maintaining and growing for the transfections. After transfection, the viability will slowly decrease to the lower 90% to 80% range by about 3-6 days post-transfection and can drop more dramatically at 7 days or longer. This is when using our recommended protocol, which does not suggest any media changes or supplementation post-transfection.

A few parameters we recommend checking to assure high culture viability are: pH at ˜7.0, no visible clumping of the cells, CO2 levels at approximately 7-8% (this should be adjusted to achieve the appropriate pH), volume of culture at 40% or less of nominal flask capacity, and incubator temperature at 37 degrees C. If you have a shaker platform setup in your incubator, the platform may generate sufficient heat to increase the temperature of the incubator. We recommend using a thermometer attached to the shaker platform. Use of baffled flasks may help, if insufficient oxygenation of the cultures is suspected. We also recommend handling and storing the culture media properly, as light exposure can result in breakdown of certain media components. If you are using FreeStyle 293 Expression Medium, supplementing with Pluronic reagent is not necessary.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.