Learn More
Aat Bioquest Pluronic F-127 Cell culture tested (10 g)
SDP
Catalog No. 502249801
Encompass_Preferred
Pluronic F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells and frequently used with dye AM esters to improve their water solubility. This catalog is for 10 g.
This item is not returnable. View return policy
Catalog No. 50-224-9801 Supplier Aat Bioquest Supplier No. 20050
Add to Cart
Add to Cart
Pluronic F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells and frequently used with dye AM esters to improve their water solubility. This catalog is for 10 g.