Learn More
Aat Bioquest Pluronic F-127 10% solution in water (10 mL)
SDP
Supplier:  Aat Bioquest 20053
Encompass_Preferred
Pluronic F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells and frequently used with dye AM esters to improve their water solubility. This catalog is for 10 mL.
Catalog No. 50-224-9802