Learn More
Bio X Cell Antibody, Clone 2.4G2, Mouse CD16/CD32, 100 MG
Supplier: Bio X Cell BP0307R100MG
The 2.4G2 monoclonal antibody reacts with mouse CD16 (FcγRIII) and CD32 (FcγRII), which are found on various immune cells. These receptors bind to the Fc portion of antibody-antigen complexes and contribute to adaptive immune responses. This antibody is commonly used to prevent non-specific binding of IgG Fc portions in immunoassays.
- Clone: 2.4G2
- Target: Mouse CD16 (FcγRIII) and CD32 (FcγRII)
- Host species: Rat
- Isotype: Rat IgG2b, κ
- Applications: In vivo Fc receptor blocking, flow cytometry, immunofluorescence
- Purification method: Protein G
- Formulation: PBS, pH 8.0
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.