Learn More
Bio X Cell Monoclonal Antibody, anti-mouse CD16/CD32, 2.4G2, 25 MG
Supplier: Bio X Cell BP0307R025MG
The 2.4G2 monoclonal antibody specifically reacts with mouse CD16 (FcγRIII) and CD32 (FcγRII), which are expressed on various immune cells and play a role in adaptive immune responses. This antibody is commonly used in flow cytometry and immunofluorescence to prevent non-specific binding of IgG Fc portions to FcγIII and FcγII receptors. Both the complete antibody and Fab fragments are effective for in vivo Fc receptor blocking.
- Clone: 2.4G2
- Reactivity: Mouse CD16 (FcγRIII) and CD32 (FcγRII)
- Isotype: Rat IgG2b, κ
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: ≤0.5EU/mg
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification method: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.