Learn More
Bio X Cell Antibody, Clone 2.4G2, CD16/CD32, 5 MG
Supplier: Bio X Cell BP0307R005MG
The InVivoPlus anti-mouse CD16/CD32 antibody is specifically designed for in vivo Fc receptor blocking applications.
- Applications: in vivo Fc receptor blocking, flow cytometry, immunofluorescence
- Host species: Rat
- Species reactivity: Mouse
- Isotype: IgG2b, κ
- Immunogen: BALB/c mouse macrophage cell line J774
- Storage conditions: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.