Learn More
Bio X Cell 2.4G2 Antibody, Mouse, Rat IgG2b, 50 MG
Supplier: Bio X Cell BP0307R050MG
The 2.4G2 monoclonal antibody specifically reacts with mouse CD16 (FcγRIII) and CD32 (FcγRII), which are expressed on various immune cells. This antibody is used in flow cytometry and immunofluorescence to prevent non-specific Fc-mediated binding and for in vivo Fc receptor blocking.
- Clone: 2.4G2
- Reactivity: mouse
- Isotype: rat IgG2b, κ
- Immunogen: BALB/c mouse macrophage cell line J774
- Formulation: PBS, pH 8.0, no stabilizers or preservatives
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification method: protein G
- Molecular weight: 150 kDa
- Storage conditions: 4°C, do not freeze
- Applications: in vivo Fc receptor blocking, flow cytometry, immunofluorescence
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.