Learn More
Precision Plus Vacuum Parts Inc Plus 530 Oil - 1 Liter
Catalog No. 501956225
Encompass_Preferred
Plus 530 in a 100% hydrocarbon, additive-free oil designed for use in large rotary vane and piston/lobe type vacuum pumps. The high viscosity and low pour point ensures ease of handling in extreme temperatures. Equivalent to an SAE 30 WT oil
Catalog No. 50-195-6225 Supplier Precision Plus Vacuum Parts Inc Supplier No. M100001
Plus 530 in a 100% hydrocarbon, additive-free oil designed for use in large rotary vane and piston/lobe type vacuum pumps. The high viscosity and low pour point ensures ease of handling in extreme temperatures. Equivalent to an SAE 30 WT oil