Learn More
Enzo Life Sciences POLYVIEW PLUS AP (anti-mouse) reagent (150tests)
SDP
Supplier:  Enzo Life Sciences ENZACC1140150
Encompass_Preferred
The POLYVIEW PLUS AP (anti-mouse) reagent is a ready-to-use, biotin-free, one-step detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the POLYVIEW PLUS AP (anti-mouse) reagent can be adapted for in situ hybridization detection. Applications: IHC ISH (in situ hybridization).
Catalog No. 50-201-3362