Learn More
Insphero Inc Akura™ PLUS Hanging Drop System - powered by InSphero
SDP
Catalog No. NC2165302
Encompass
The world-famous and unique hanging drop plate in 96-well format with automation-compatible top-loading.1. Loading of cell suspension from the top through unique SureDrop™ inlet engineered for highly uniform and stable hanging drops2. Gravity-assisted formation of uniform spheroid/organoid morphology and functionality in long-term culture, results in one spheroid per well3. Suitable for many cell types, including cells which don't normally form spheroids or organoids on ultra-low attachment surfaces Contains 10 Akura™ PLUS Hanging Drop Plates and 10 Akura™ 96 Spheroid Plates, individually wrapped, made of polystyrene and cyclo olefin polymer, sterile, includes lid. For further information, please visit www.insphero.com
This item is not returnable. View return policy
Catalog No. NC2165302 Supplier Insphero Inc Supplier No. CSPF24
The world-famous and unique hanging drop plate in 96-well format with automation-compatible top-loading.1. Loading of cell suspension from the top through unique SureDrop™ inlet engineered for highly uniform and stable hanging drops2. Gravity-assisted formation of uniform spheroid/organoid morphology and functionality in long-term culture, results in one spheroid per well3. Suitable for many cell types, including cells which don't normally form spheroids or organoids on ultra-low attachment surfaces Contains 10 Akura™ PLUS Hanging Drop Plates and 10 Akura™ 96 Spheroid Plates, individually wrapped, made of polystyrene and cyclo olefin polymer, sterile, includes lid. For further information, please visit www.insphero.com