Learn More
Enzo Life Sciences POLYVIEW PLUS HRP-DAB (Anti-Mouse) Kit (150 tests)
SDP
Supplier:  Enzo Life Sciences ENZKIT1600150
Encompass_Preferred
The POLYVIEW PLUS HRP-DAB (Anti-Mouse) Kit is intended for both IHC and ISH staining. The kit is a non-biotin one step detection system suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections. This kit can be used with blood smears, cytosmears, and cell preparations. Assay Time: < 2 hours. Applications: IHC ISH (in situ hybridization).
Catalog No. 50-201-3851