Learn More
Meso Scale Discovery V-PLEX Plus Human IFN-γ Kit, 5 plates, 96-well, 0.37 pg/mL LLOD, dynamic range 0.37 - 938 pg/mL, suitable for serum, plasma, urine, cell culture supernatant, Mouse Monoclonal antibodies
SDP
Catalog No. NC3240399
Encompass
The MESO SCALE DISCOVERY V-PLEX Plus Human IFN-γ Kit is a high-performance single-analyte immunoassay designed to quantitatively measure human Interferon-gamma in various biological samples. This kit provides exceptional sensitivity and accuracy, as it belongs to the analytically validated V-PLEX platform, ensuring reliable results across different research applications. The kit includes essential components for easy setup and is ideal for assays requiring high sensitivity and broad dynamic range.
  • Designed for quantitative measurement of human IFN-γ
  • Supports various sample types including serum and plasma
  • Offers sensitivity down to 0.37 pg/mL
  • Features a dynamic range from 0.37 to 938 pg/mL
  • Includes multiple plates for high-throughput testing
  • Utilizes mouse monoclonal antibodies for both capture and detection
  • Validated with MESO SECTOR and QuickPlex instruments
  • For research use only
Catalog No. NC3240399 Supplier Meso Scale Discovery Supplier No. K151QOG2
The MESO SCALE DISCOVERY V-PLEX Plus Human IFN-γ Kit is a high-performance single-analyte immunoassay designed to quantitatively measure human Interferon-gamma in various biological samples. This kit provides exceptional sensitivity and accuracy, as it belongs to the analytically validated V-PLEX platform, ensuring reliable results across different research applications. The kit includes essential components for easy setup and is ideal for assays requiring high sensitivity and broad dynamic range.
  • Designed for quantitative measurement of human IFN-γ
  • Supports various sample types including serum and plasma
  • Offers sensitivity down to 0.37 pg/mL
  • Features a dynamic range from 0.37 to 938 pg/mL
  • Includes multiple plates for high-throughput testing
  • Utilizes mouse monoclonal antibodies for both capture and detection
  • Validated with MESO SECTOR and QuickPlex instruments
  • For research use only