Learn More
Bio X Cell Monoclonal Antibody, Mouse CD20, MB20-11, 25 MG
Supplier: Bio X Cell BP0356R025MG
The MB20-11 monoclonal antibody reacts with mouse CD20, a B cell-specific transmembrane protein involved in intracellular calcium regulation and B cell activation. This antibody is designed for rapid and effective B cell depletion via antibody-dependent cellular cytotoxicity (ADCC).
- Reacts with mouse CD20
- Isotype: mouse IgG2c, κ
- Immunogen: mouse CD20-GFP transfected 300.19 cells
- Applications include in vivo B cell depletion and Western blot
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level ≤0.5EU/mg
- Aggregation <5%
- Purity ≥95%
- Sterilized by 0.2 μm filtration
- Produced from cell culture supernatant in an animal-free facility
- Purified by Protein A
- Molecular weight: 150 kDa
- Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.