Learn More
Bio X Cell InVivoPlus Mouse CD20, MB20-11, IgG2c, 50 MG
Supplier: Bio X Cell BP0356R050MG
The MB20-11 monoclonal antibody reacts with mouse CD20, a B cell-specific transmembrane protein involved in intracellular calcium regulation and B cell activation. This antibody effectively depletes B cells via antibody-dependent cellular cytotoxicity (ADCC).
- Clone: MB20-11
- Target: Mouse CD20
- Host species: Mouse
- Isotype: Mouse IgG2c, κ
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: ≤0.5EU/mg
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Sterility: 0.2 μm filtration
- Applications: in vivo B cell depletion, Western blot
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.