Learn More
Bio X Cell Antibody, Clone MIAP410, Mouse/Human/Rat CD47, 100 MG
Supplier: Bio X Cell BP0283R100MG
The MIAP410 monoclonal antibody reacts with mouse CD47, an integrin-associated protein (IAP) ubiquitously expressed by hematopoietic and non-hematopoietic cells. CD47 is involved in various cellular processes including apoptosis, proliferation, adhesion, and migration, and plays a role in immune and angiogenic responses. The MIAP410 antibody has demonstrated neutralization of CD47 both in vivo and in vitro.
- Reacts with mouse, human, and rat CD47
- Isotype: Mouse IgG1, κ
- Immunogen: Purified human placental CD47
- Reported applications: In vivo CD47 blockade, in vitro CD47 blocking, immunofluorescence
- Purity: ≥95%, determined by SDS-PAGE
- Endotoxin: ≤0.5EU/mg
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.