Learn More
Bio X Cell InVivoPlus Antibody, CD47, Clone MIAP410, 25 MG
Supplier: Bio X Cell BP0283R025MG
The MIAP410 monoclonal antibody targets CD47 (integrin-associated protein), a ∼50 kDa glycosylated protein involved in cellular processes and immune responses. Often overexpressed in tumor cells, CD47 is a therapeutic target. This antibody effectively neutralizes CD47 both in vivo and in vitro.
- Neutralizes CD47 in vivo and in vitro
- Reactivity against mouse, human, and rat CD47
- Purified by Protein G from animal-free facility
- Purity ≥95% (SDS-PAGE), aggregation <5% (SEC)
- Endotoxin ≤0.5 EU/mg (≤0.0005 EU/μg)
- Sterile filtered (0.2 μm) for high quality
- Suitable for immunofluorescence applications
- Formulated in PBS, pH 7.0; no stabilizers or preservatives
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.