Learn More
Bio X Cell Antibody, MIAP410, Mouse, Human, Unconjugated, 5 MG
Supplier: Bio X Cell BP0283R005MG
The MIAP410 monoclonal antibody targets mouse CD47, also known as integrin-associated protein (IAP). CD47 is a ˜50 kDa glycosylated transmembrane protein expressed ubiquitously on hematopoietic and non-hematopoietic cells. It participates in cellular processes like apoptosis, proliferation, adhesion, and migration, crucial for immune and angiogenic responses.
- Clone: MIAP410
- Reactivities: Mouse, Human, Rat
- Isotype: Mouse IgG1, κ
- Unconjugated
- Immunogen: Purified human placental CD47
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg)
- Aggregation: <5%
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store antibody solution at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.