Learn More
Bio X Cell Antibody, anti-CD47, MIAP410, Mouse IgG1, 50 MG
Supplier: Bio X Cell BP0283R050MG
The MIAP410 monoclonal antibody targets mouse CD47 (IAP), a ~50 kDa glycosylated transmembrane protein involved in immune and angiogenic responses. Overexpressed in many tumor cells, anti-CD47 antibodies like MIAP410 are studied for cancer treatment, neutralizing CD47 in vivo and in vitro.
- Isotype: Mouse IgG1, κ
- Immunogen: Purified human placental CD47
- Applications: In vivo CD47 blockade, in vitro CD47 blocking, immunofluorescence
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: ≤0.5EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Storage: Store at 4°C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.