Learn More
Apexbio Technology LLC (+)-MK 801 Maleate 77086-22-7 10mg
SDP
Catalog No. 5000062310
Encompass_Preferred
( )-MK 801 Maleate is a non-competitive antagonist of the N-methyl-D-aspartate (NMDA) receptor exhibiting a Ki value of approximately 30 5 nM By blocking the NMDA receptor-mediated ion channel MK 801 disrupts glutamate-associated excitatory neurotransmission In experimental models MK 801 demonstrates anticonvulsant and anxiolytic properties through modulation of NMDA receptor activity This compound readily penetrates the central nervous system binding reversibly and region-specifically to cortical and hippocampal membranes MK 801 effectively inhibits NMDA-triggered depolarization responses in neuronal preparations making it applicable in studies focused on epilepsy excitotoxicity and neuronal signaling pathways
Catalog No. 50-000-62310 Supplier Apexbio Technology LLC Supplier No. A889610
( )-MK 801 Maleate is a non-competitive antagonist of the N-methyl-D-aspartate (NMDA) receptor exhibiting a Ki value of approximately 30 5 nM By blocking the NMDA receptor-mediated ion channel MK 801 disrupts glutamate-associated excitatory neurotransmission In experimental models MK 801 demonstrates anticonvulsant and anxiolytic properties through modulation of NMDA receptor activity This compound readily penetrates the central nervous system binding reversibly and region-specifically to cortical and hippocampal membranes MK 801 effectively inhibits NMDA-triggered depolarization responses in neuronal preparations making it applicable in studies focused on epilepsy excitotoxicity and neuronal signaling pathways