Learn More
MSC Vermont Gage Plug Gage, Black Guard, 0.0250 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000513903
Encompass
This Class ZZ plus plug gage is a high-precision measurement tool featuring a 0.0250 in. diameter and 2 in. overall length. Engineered for reliable inspection, this gage is constructed from tool steel with a durable Black Guard™ finish to provide long-lasting performance in demanding metalworking and inspection environments.
  • Tool steel construction with 60/65 Rc hardness
  • Surface finish of 10 microinch or better
  • Go (plus) tolerance of +0.0002 / -0.0000 in.
  • NIST traceable calibration
  • Certificate of accuracy included
  • Precision cylindrical design
Catalog No. 50-005-13903 Supplier MSC Supplier No. 51462497
This Class ZZ plus plug gage is a high-precision measurement tool featuring a 0.0250 in. diameter and 2 in. overall length. Engineered for reliable inspection, this gage is constructed from tool steel with a durable Black Guard™ finish to provide long-lasting performance in demanding metalworking and inspection environments.
  • Tool steel construction with 60/65 Rc hardness
  • Surface finish of 10 microinch or better
  • Go (plus) tolerance of +0.0002 / -0.0000 in.
  • NIST traceable calibration
  • Certificate of accuracy included
  • Precision cylindrical design