Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0455 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000513072
Encompass
This Class ZZ plug gage is designed for precise measurement applications, including Go/No Go gaging and the inspection of hole sizes. Manufactured from 52100 tool steel and treated to a hardness of 60 to 62 Rc, this gage provides durability and long-lasting accuracy. Each unit features a black oxide finish and is centerless lapped to a surface finish of 10 microinch or better.
  • NIST traceable with included certificate of accuracy
  • Measures hole sizes and calculates distances between holes and slot widths
  • Features plus tolerance of 0.0002 in.
  • Diameter size is electro etched on each piece
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Constructed to ASME B89.1 standards
Catalog No. 50-005-13072 Supplier MSC Supplier No. 72789670
This Class ZZ plug gage is designed for precise measurement applications, including Go/No Go gaging and the inspection of hole sizes. Manufactured from 52100 tool steel and treated to a hardness of 60 to 62 Rc, this gage provides durability and long-lasting accuracy. Each unit features a black oxide finish and is centerless lapped to a surface finish of 10 microinch or better.
  • NIST traceable with included certificate of accuracy
  • Measures hole sizes and calculates distances between holes and slot widths
  • Features plus tolerance of 0.0002 in.
  • Diameter size is electro etched on each piece
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Constructed to ASME B89.1 standards