Learn More
Abcam (±)-Acetylcarnitine chloride, cholinergic agonist, 250MG
Catalog No. 502706883
Encompass_Preferred
MW 239.69 Da, Purity >98%. Important intermediate in lipid metabolism. Weak cholinergic agonist. Metabolic and antioxidant modulator, with ability to cross the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6883 Supplier Abcam Supplier No. AB141121250MG
MW 239.69 Da, Purity >98%. Important intermediate in lipid metabolism. Weak cholinergic agonist. Metabolic and antioxidant modulator, with ability to cross the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement