Learn More
Abcam (±)-Clopidogrel sulfate, P2Y12 antagonist, 10MG
Catalog No. 502707627
Encompass_Preferred
MW 419.89 g/mol, Purity >99%. Potent, selective P2Y₁₂ antagonist. Potent antithrombotic drug that inhibits ADP-induced platelet aggregation. Clopidogrel is not active in vitro and is the precursor to an active metabolite formed in the liver.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7627 Supplier Abcam Supplier No. AB12038710MG
MW 419.89 g/mol, Purity >99%. Potent, selective P2Y₁₂ antagonist. Potent antithrombotic drug that inhibits ADP-induced platelet aggregation. Clopidogrel is not active in vitro and is the precursor to an active metabolite formed in the liver.
The product is subject to the following: Abcam Restricted Use Statement