Learn More
Abcam (±)-Miconazole nitrate, Imidazole antifungal agent, 1G
Catalog No. 502706813
Encompass_Preferred
MW 479.1 Da, Purity >99%. Imidazole antifungal agent. Inhibits fungal membrane ergosterol biosynthesis. Increases reactive oxygen species production.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6813 Supplier Abcam Supplier No. AB1411951G
MW 479.1 Da, Purity >99%. Imidazole antifungal agent. Inhibits fungal membrane ergosterol biosynthesis. Increases reactive oxygen species production.
The product is subject to the following: Abcam Restricted Use Statement