Learn More
Abcam (±)-Miconazole nitrate, Imidazole antifungal agent, 5G
Catalog No. 502706812
Encompass_Preferred
MW 479.1 Da, Purity >99%. Imidazole antifungal agent. Inhibits fungal membrane ergosterol biosynthesis. Increases reactive oxygen species production.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6812 Supplier Abcam Supplier No. AB1411955G
MW 479.1 Da, Purity >99%. Imidazole antifungal agent. Inhibits fungal membrane ergosterol biosynthesis. Increases reactive oxygen species production.
The product is subject to the following: Abcam Restricted Use Statement