Learn More
Abcam (±)-Naringenin, Antio x idant, 50MG
Catalog No. 502707087
Encompass_Preferred
MW 272.26 g/mol, Purity >98%. Citrus flavonoid. Antioxidant, anti-inflammatory. Affects various signaling pathways in vitro and in vivo. Crosses the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7087 Supplier Abcam Supplier No. AB12095850MG
MW 272.26 g/mol, Purity >98%. Citrus flavonoid. Antioxidant, anti-inflammatory. Affects various signaling pathways in vitro and in vivo. Crosses the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement