Learn More
Abcam (±)-Propionylcarnitine chloride, homolog of acetylcarnitine chloride, 50MG
Supplier:  Abcam AB14112350MG
Encompass_Preferred
MW 252.73 g/mol, Purity >98%. Naturally occuring homolog of acetylcarnitine chloride (ab141121). Essential for mitochondrial fatty acid transport and high-energy phosphate exchange.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6881