Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Plasmalemma vesicle-associated protein (PLVAP), human recombinant (HEK293, His tag) | ≥95.0% | 100UG
SDP

Catalog No. 5000175607
Encompass_Preferred

Recombinant human plasmalemma vesicle-associated protein (PLVAP) is supplied as a lyophilized, HEK293-expressed His-tagged protein for research on endothelial barrier function and microvascular permeability. The material is provided with low endotoxin and guidance for reconstitution and storage to support biochemical and cell-based studies.

  • Species: human.
  • Source: HEK293 expression system.
  • Tag: His tag for purification.
  • Purity: ≥95.0% as determined by Tris-Bis PAGE.
  • Molecular weight: 50-70 kDa.
  • Formulation: lyophilized from PBS with 200 mM l-arginine and 8% trehalose.
  • Endotoxin: <1 EU/μg.
  • Storage: store lyophilized at -20°C; aliquot and freeze after reconstitution.

Catalog No. 50-001-75607 Supplier Medchemexpress LLC Supplier No. HYP700879100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human plasmalemma vesicle-associated protein (PLVAP) is supplied as a lyophilized, HEK293-expressed His-tagged protein for research on endothelial barrier function and microvascular permeability. The material is provided with low endotoxin and guidance for reconstitution and storage to support biochemical and cell-based studies.

  • Species: human.
  • Source: HEK293 expression system.
  • Tag: His tag for purification.
  • Purity: ≥95.0% as determined by Tris-Bis PAGE.
  • Molecular weight: 50-70 kDa.
  • Formulation: lyophilized from PBS with 200 mM l-arginine and 8% trehalose.
  • Endotoxin: <1 EU/μg.
  • Storage: store lyophilized at -20°C; aliquot and freeze after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.